Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD6 SIRNA Assays ›
Gene Symbol:
ANKRD6
Gene Name:
ankyrin repeat domain 6
Gene Aliases:
-
Chromosome Location:
Chr.6: 89433178 - 89633834 on Build GRCh38
UniGene ID:
Hs.702213
Species:
Human
siRNA ID
HSS117811
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242809.1
NP_001229738.1
16
2457
NM_001242811.1
NP_001229740.1
16
2840
NM_001242813.1
NP_001229742.1
15
2352
NM_001242814.1
NP_001229743.1
13
2063
NM_014942.4
NP_055757.3
16
2442
XM_005248679.3
XP_005248736.1
15
2325
XM_005248680.3
XP_005248737.1
17
4614
XM_005248681.2
XP_005248738.1
16
2558
XM_005248682.3
XP_005248739.1
14
2247
XM_005248683.3
XP_005248740.1
13
2127
XM_005248684.3
XP_005248741.1
10
1845
XM_011535596.2
XP_011533898.1
14
2226
XM_011535597.2
XP_011533899.1
14
2226
XM_011535598.2
XP_011533900.1
15
2327
XM_011535599.2
XP_011533901.1
15
2327
XM_011535600.2
XP_011533902.1
14
2234
XM_017010493.1
XP_016865982.1
14
2220
XM_017010494.1
XP_016865983.1
14
2211
XM_017010495.1
XP_016865984.1
15
2327
XM_017010496.1
XP_016865985.1
15
2491
XM_017010497.1
XP_016865986.1
14
2145
XM_017010498.1
XP_016865987.1
15
2306
XM_017010499.1
XP_016865988.1
13
2121
XM_017010500.1
XP_016865989.1
13
2112
XM_017010501.1
XP_016865990.1
14
2228
XM_017010502.1
XP_016865991.1
14
3909
XM_017010503.1
XP_016865992.1
14
3913
XM_017010504.1
XP_016865993.1
13
2055
XM_017010505.1
XP_016865994.1
14
2222
XM_017010506.1
XP_016865995.1
13
2046
XM_017010507.1
XP_016865996.1
13
2046
XM_017010508.1
XP_016865997.1
13
3805
XM_017010509.1
XP_016865998.1
12
1956
XM_017010510.1
XP_016865999.1
12
2011
XM_017010511.1
XP_016866000.1
12
3714
XM_017010512.1
XP_016866001.1
11
1946
XM_017010513.1
XP_016866002.1
11
2078
XM_017010514.1
XP_016866003.1
10
1847
XM_017010515.1
XP_016866004.1
16
2342
GenBank mRNA
AB023174.1
-
15
2179
AK074189.1
-
8
1460
AK092235.1
-
7
2032
AK096867.1
-
13
1925
AK295731.1
-
16
2757
AK307792.1
-
16
2426
AL117504.1
-
13
1858
BC001078.2
-
4
894
BC042173.1
-
16
2407
5454509
GU727626.1
-
4
790
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
kinase inhibitor
kinase modulator
non-receptor serine/threonine protein kinase
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
positive regulation of JNK cascade
negative regulation of canonical Wnt signaling pathway
positive regulation of Wnt signaling pathway, planar cell polarity pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0