Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD6 SIRNA Assays ›
Gene Symbol:
ANKRD6
Gene Name:
ankyrin repeat domain 6
Gene Aliases:
-
Chromosome Location:
Chr.6: 89433178 - 89633834 on Build GRCh38
UniGene ID:
Hs.702213
Species:
Human
siRNA ID
HSS117813
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242809.1
NP_001229738.1
8
1036
NM_001242811.1
NP_001229740.1
8
1419
NM_001242813.1
NP_001229742.1
8
1036
NM_001242814.1
NP_001229743.1
6
735
NM_014942.4
NP_055757.3
8
1036
XM_005248679.3
XP_005248736.1
7
904
XM_005248680.3
XP_005248737.1
9
3208
XM_005248681.2
XP_005248738.1
8
1137
XM_005248682.3
XP_005248739.1
7
904
XM_005248683.3
XP_005248740.1
5
706
XM_011535596.2
XP_011533898.1
6
805
XM_011535597.2
XP_011533899.1
6
805
XM_011535598.2
XP_011533900.1
7
906
XM_011535599.2
XP_011533901.1
7
906
XM_011535600.2
XP_011533902.1
7
906
XM_017010493.1
XP_016865982.1
7
904
XM_017010494.1
XP_016865983.1
6
805
XM_017010495.1
XP_016865984.1
7
906
XM_017010496.1
XP_016865985.1
7
1070
XM_017010497.1
XP_016865986.1
6
739
XM_017010498.1
XP_016865987.1
8
1005
XM_017010499.1
XP_016865988.1
6
805
XM_017010500.1
XP_016865989.1
5
706
XM_017010501.1
XP_016865990.1
6
807
XM_017010502.1
XP_016865991.1
6
2488
XM_017010503.1
XP_016865992.1
6
2492
XM_017010504.1
XP_016865993.1
6
739
XM_017010505.1
XP_016865994.1
7
906
XM_017010506.1
XP_016865995.1
5
640
XM_017010507.1
XP_016865996.1
5
640
XM_017010508.1
XP_016865997.1
5
2384
XM_017010509.1
XP_016865998.1
5
640
XM_017010510.1
XP_016865999.1
5
695
XM_017010511.1
XP_016866000.1
5
2398
XM_017010515.1
XP_016866004.1
7
838
GenBank mRNA
AB023174.1
-
8
863
AK096867.1
-
6
597
AK295731.1
-
8
1336
AK307792.1
-
8
1005
AL117504.1
-
5
437
BC042173.1
-
8
1001
5454509
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
kinase inhibitor
kinase modulator
non-receptor serine/threonine protein kinase
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
positive regulation of JNK cascade
negative regulation of canonical Wnt signaling pathway
positive regulation of Wnt signaling pathway, planar cell polarity pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline