Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143854.1 | NP_001137326.1 | 12 | 1335 | ||
NM_014954.3 | NP_055769.2 | 11 | 1323 | |||
XM_011538048.1 | XP_011536350.1 | 12 | 1139 | |||
XM_017019034.1 | XP_016874523.1 | 4 | 593 | |||
GenBank mRNA | AB023202.1 | - | 12 | 1256 | ||
AK295696.1 | - | 12 | 1289 | |||
AK296296.1 | - | 10 | 874 | |||
AK316112.1 | - | 12 | 1170 | |||
BC002605.2 | - | 10 | 1061 | |||
BC009843.2 | - | 10 | 984 | |||
BC017259.2 | - | 11 | 1130 | 3510158 | ||
BX647281.1 | - | 4 | 1929 |