Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP164 SIRNA Assays ›
Gene Symbol:
CEP164
Gene Name:
centrosomal protein 164
Gene Aliases:
NPHP15
Chromosome Location:
Chr.11: 117316346 - 117413266 on Build GRCh38
UniGene ID:
Hs.504009
Species:
Human
siRNA ID
HSS117847
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP164
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271933.1
NP_001258862.1
14
1864
NM_014956.4
NP_055771.4
15
1921
XM_005271453.1
XP_005271510.1
16
3878
XM_005271456.1
XP_005271513.1
15
2192
XM_005271457.1
XP_005271514.1
15
2201
XM_006718788.1
XP_006718851.1
16
3887
XM_006718794.1
XP_006718857.1
14
2114
XM_011542674.1
XP_011540976.1
16
3938
XM_017017364.1
XP_016872853.1
16
3805
XM_017017365.1
XP_016872854.1
16
3957
XM_017017366.1
XP_016872855.1
16
3740
XM_017017367.1
XP_016872856.1
15
3866
XM_017017368.1
XP_016872857.1
16
3938
XM_017017369.1
XP_016872858.1
16
3938
XM_017017370.1
XP_016872859.1
16
3938
XM_017017371.1
XP_016872860.1
15
3860
XM_017017372.1
XP_016872861.1
15
3800
XM_017017373.1
XP_016872862.1
15
3738
XM_017017374.1
XP_016872863.1
15
3533
XM_017017375.1
XP_016872864.1
14
3662
XM_017017376.1
XP_016872865.1
15
3791
XM_017017377.1
XP_016872866.1
17
3897
XM_017017379.1
XP_016872868.1
15
2252
XM_017017380.1
XP_016872869.1
14
1914
XM_017017381.1
XP_016872870.1
14
2174
XM_017017382.1
XP_016872871.1
13
1836
XM_017017383.1
XP_016872872.1
14
2103
XM_017017384.1
XP_016872873.1
14
1902
XM_017017385.1
XP_016872874.1
12
1658
XM_017017386.1
XP_016872875.1
12
1818
GenBank mRNA
AB028975.1
-
14
2135
AK125995.1
-
10
1890
AK303067.1
-
13
1775
AL137265.1
-
8
1098
AL833532.1
-
9
1305
BX647583.1
-
1
2656
BX648682.1
-
15
1921
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
DNA repair
mitotic nuclear division
cilium assembly
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0