Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP164 SIRNA Assays ›
Gene Symbol:
CEP164
Gene Name:
centrosomal protein 164
Gene Aliases:
NPHP15
Chromosome Location:
Chr.11: 117316346 - 117413266 on Build GRCh38
UniGene ID:
Hs.504009
Species:
Human
siRNA ID
HSS117849
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP164
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271933.1
NP_001258862.1
26
3429
NM_014956.4
NP_055771.4
27
3486
XM_005271453.1
XP_005271510.1
28
5443
XM_005271456.1
XP_005271513.1
27
3757
XM_005271457.1
XP_005271514.1
27
3766
XM_006718788.1
XP_006718851.1
28
5452
XM_006718794.1
XP_006718857.1
26
3679
XM_011542674.1
XP_011540976.1
28
5503
XM_017017364.1
XP_016872853.1
28
5370
XM_017017365.1
XP_016872854.1
28
5522
XM_017017366.1
XP_016872855.1
28
5305
XM_017017367.1
XP_016872856.1
27
5431
XM_017017368.1
XP_016872857.1
28
5500
XM_017017369.1
XP_016872858.1
28
5503
XM_017017370.1
XP_016872859.1
28
5500
XM_017017371.1
XP_016872860.1
27
5425
XM_017017372.1
XP_016872861.1
27
5365
XM_017017373.1
XP_016872862.1
27
5303
XM_017017374.1
XP_016872863.1
27
5098
XM_017017375.1
XP_016872864.1
26
5227
XM_017017376.1
XP_016872865.1
27
5356
XM_017017377.1
XP_016872866.1
29
5462
XM_017017378.1
XP_016872867.1
26
5146
XM_017017379.1
XP_016872868.1
27
3817
XM_017017380.1
XP_016872869.1
26
3479
XM_017017381.1
XP_016872870.1
26
3739
XM_017017382.1
XP_016872871.1
25
3401
XM_017017383.1
XP_016872872.1
26
3668
XM_017017384.1
XP_016872873.1
26
3467
XM_017017385.1
XP_016872874.1
24
3223
XM_017017386.1
XP_016872875.1
24
3383
GenBank mRNA
AB028975.1
-
26
3700
AK001412.1
-
3
230
AK125995.1
-
21
3574
AL117632.1
-
8
748
AL137265.1
-
20
2663
BX647583.1
-
13
4221
BX648682.1
-
27
3486
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
DNA repair
mitotic nuclear division
cilium assembly
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-cq5mm:80/10.239.166.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0