Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014282.3 | NP_055097.2 | 6 | 1068 | ||
XM_005251812.3 | XP_005251869.1 | 6 | 899 | |||
XM_017014484.1 | XP_016869973.1 | 6 | 730 | |||
GenBank mRNA | AF241831.1 | - | 6 | 1018 | ||
AK000610.1 | - | 1 | 109 | |||
AK025144.1 | - | 4 | 415 | |||
AK055161.1 | - | 1 | 1426 | |||
AK091242.1 | - | 1 | 2301 | |||
BC018788.1 | - | 3 | 707 | 3502216 | ||
U77327.1 | - | 5 | 601 |