Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF25 SIRNA Assays ›
Gene Symbol:
TCF25
Gene Name:
transcription factor 25
Gene Aliases:
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049
Chromosome Location:
Chr.16: 89873583 - 89911384 on Build GRCh38
UniGene ID:
Hs.415342
Species:
Human
siRNA ID
HSS117977
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF25
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014972.2
NP_055787.1
12
1348
XM_005256297.2
XP_005256354.1
12
1342
XM_005256298.2
XP_005256355.1
12
1342
XM_005256299.2
XP_005256356.1
12
1297
XM_011522956.2
XP_011521258.1
12
1357
XM_011522957.2
XP_011521259.1
12
1357
XM_011522958.2
XP_011521260.1
12
1312
XM_011522959.2
XP_011521261.1
11
1291
XM_011522960.2
XP_011521262.1
12
1357
XM_011522961.2
XP_011521263.1
13
1526
XM_017023047.1
XP_016878536.1
11
1276
XM_017023048.1
XP_016878537.1
12
1297
XM_017023049.1
XP_016878538.1
12
1312
XM_017023050.1
XP_016878539.1
12
1297
XM_017023051.1
XP_016878540.1
11
1276
XM_017023052.1
XP_016878541.1
11
1231
XM_017023053.1
XP_016878542.1
13
1964
XM_017023054.1
XP_016878543.1
13
1511
XM_017023055.1
XP_016878544.1
13
1466
XM_017023056.1
XP_016878545.1
13
1518
XM_017023057.1
XP_016878546.1
13
1949
XM_017023058.1
XP_016878547.1
12
1883
GenBank mRNA
AB028972.1
-
10
889
AF130092.1
-
9
850
AF322111.1
-
12
1362
BC000959.1
-
12
1271
3445789
BC009349.2
-
12
1293
4136178
BC011884.2
-
12
1322
3678670
DQ321703.1
-
12
1362
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
heart development
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0