Search Thermo Fisher Scientific
Gene Symbol: PDS5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015032.3 | NP_055847.1 | 17, 18 | 2040 | ||
XM_005266298.4 | XP_005266355.1 | 17, 18 | 2035 | |||
XM_011534999.2 | XP_011533301.1 | 17, 18 | 2035 | |||
XM_011535000.2 | XP_011533302.1 | 17, 18 | 2035 | |||
XM_011535002.2 | XP_011533304.1 | 10, 11 | 1137 | |||
XM_017020448.1 | XP_016875937.1 | 17, 18 | 2182 | |||
XM_017020449.1 | XP_016875938.1 | 17, 18 | 1950 | |||
XM_017020450.1 | XP_016875939.1 | 17, 18 | 1950 | |||
XM_017020451.1 | XP_016875940.1 | 17, 18 | 1950 | |||
XM_017020452.1 | XP_016875941.1 | 17, 18 | 2006 | |||
XM_017020453.1 | XP_016875942.1 | 17, 18 | 2035 | |||
GenBank mRNA | AB023196.2 | - | 17, 18 | 1962 | ||
AK299340.1 | - | 17, 18 | 1916 | |||
AL137201.1 | - | 17, 18 | 1980 | |||
U50533.1 | - | 7, 8 | 791 | |||
U95825.2 | - | 17, 18 | 1919 |