Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015092.4 | NP_055907.3 | 43 | 7325 | ||
XM_005255182.3 | XP_005255239.1 | 43 | 7338 | |||
XM_005255183.3 | XP_005255240.1 | 44 | 7396 | |||
XM_005255184.3 | XP_005255241.1 | 42 | 7182 | |||
XM_011545769.2 | XP_011544071.1 | 43 | 7338 | |||
XM_011545770.1 | XP_011544072.1 | 31 | 5135 | |||
XM_017023066.1 | XP_016878555.1 | 43 | 7148 | |||
XM_017023067.1 | XP_016878556.1 | 31 | 5198 | |||
GenBank mRNA | AB007881.2 | - | 11 | 1943 | ||
AB061371.1 | - | 44 | 7277 | |||
AF186377.1 | - | 18 | 3306 | |||
AF395444.1 | - | 41 | 6632 | |||
AK074359.1 | - | 10 | 1804 | |||
AL833298.1 | - | 4 | 664 | |||
AY014957.1 | - | 31 | 5204 |