Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164211.1 | NP_001157683.1 | 4 | 861 | ||
NM_001164213.1 | NP_001157685.1 | 4 | 861 | |||
NM_015116.2 | NP_055931.1 | 4 | 861 | |||
XM_017020483.1 | XP_016875972.1 | 4 | 884 | |||
XM_017020484.1 | XP_016875973.1 | 3 | 609 | |||
GenBank mRNA | AB023233.1 | - | 4 | 822 | ||
AK292898.1 | - | 4 | 829 | |||
AK309937.1 | - | 4 | 852 | |||
AY050631.1 | - | 4 | 854 | |||
AY050632.1 | - | 4 | 854 | |||
BC112937.1 | - | 4 | 773 | 5204613 | ||
BC117472.1 | - | 4 | 762 | 40126023 | ||
BC143883.1 | - | 4 | 762 | 9052399 |