Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FCHO1 SIRNA Assays ›
Gene Symbol:
FCHO1
Gene Name:
FCH domain only 1
Gene Aliases:
-
Chromosome Location:
Chr.19: 17747718 - 17788568 on Build GRCh38
UniGene ID:
Hs.96485
Species:
Human
siRNA ID
HSS118257
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FCHO1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161357.1
NP_001154829.1
8, 9
766
NM_001161358.1
NP_001154830.1
7, 8
676
NM_001161359.1
NP_001154831.1
6, 7
606
NM_015122.2
NP_055937.1
8, 9
770
XM_005259831.3
XP_005259888.1
7, 8
851
XM_006722701.3
XP_006722764.1
7, 8
774
XM_006722702.3
XP_006722765.1
7, 8
851
XM_011527824.2
XP_011526126.1
8, 9
766
XM_011527825.2
XP_011526127.1
8, 9
755
XM_011527826.2
XP_011526128.1
8, 9
738
XM_011527827.2
XP_011526129.1
6, 7
573
XM_011527828.2
XP_011526130.1
7, 8
669
XM_017026519.1
XP_016882008.1
8, 9
836
XM_017026520.1
XP_016882009.1
8, 9
819
XM_017026521.1
XP_016882010.1
7, 8
770
XM_017026522.1
XP_016882011.1
8, 9
766
XM_017026523.1
XP_016882012.1
8, 9
836
XM_017026524.1
XP_016882013.1
7, 8
851
XM_017026525.1
XP_016882014.1
7, 8
649
XM_017026526.1
XP_016882015.1
8, 9
766
XM_017026527.1
XP_016882016.1
7, 8
774
XM_017026528.1
XP_016882017.1
8, 9
836
GenBank mRNA
AB006628.1
-
5, 6
540
AK291410.1
-
7, 8
632
AK303623.1
-
4, 5
468
AK309403.1
-
4, 5
519
AK316458.1
-
6, 7
577
BC028021.1
-
8, 9
711
5218319
BC041130.1
-
8, 9
720
5757146
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
clathrin coat assembly
clathrin-mediated endocytosis
protein binding
AP-2 adaptor complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0