Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319100.1 | NP_001306029.1 | 3 | 1250 | ||
NM_001319101.1 | NP_001306030.1 | 3 | 1254 | |||
NM_015147.2 | NP_055962.2 | 3 | 1254 | |||
NR_134966.1 | 3 | 1254 | ||||
XM_005264219.3 | XP_005264276.1 | 3 | 1243 | |||
XM_017003633.1 | XP_016859122.1 | 3 | 1223 | |||
GenBank mRNA | AB011154.2 | - | 3 | 1254 | ||
AK000856.1 | - | 3 | 1199 | |||
AK093846.1 | - | 1 | 620 | |||
AK094069.1 | - | 1 | 1408 | |||
AK299373.1 | - | 3 | 1195 | |||
AK301173.1 | - | 4 | 1161 | |||
BC002982.1 | - | 3 | 1150 | 3543243 | ||
BC004873.2 | - | 3 | 1150 | 3678131 | ||
BC030534.1 | - | 3 | 1154 | |||
BX648774.1 | - | 2 | 757 |