Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319100.1 | NP_001306029.1 | 2, 3 | 574 | ||
NM_001319101.1 | NP_001306030.1 | 2, 3 | 578 | |||
NM_015147.2 | NP_055962.2 | 2, 3 | 578 | |||
NR_134966.1 | 2, 3 | 578 | ||||
XM_005264219.3 | XP_005264276.1 | 2, 3 | 567 | |||
XM_017003633.1 | XP_016859122.1 | 2, 3 | 547 | |||
GenBank mRNA | AB011154.2 | - | 2, 3 | 578 | ||
AK299373.1 | - | 2, 3 | 519 | |||
AK301173.1 | - | 3, 4 | 485 | |||
AK304110.1 | - | 2, 3 | 523 | |||
BC002982.1 | - | 2, 3 | 474 | 3543243 | ||
BC004873.2 | - | 2, 3 | 474 | 3678131 | ||
BC030534.1 | - | 2, 3 | 478 |