Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270940.1 | NP_001257869.1 | 4 | 854 | ||
NM_015171.3 | NP_055986.1 | 3 | 718 | |||
XM_011545772.2 | XP_011544074.1 | 3 | 387 | |||
GenBank mRNA | AB002368.3 | - | 3 | 669 | ||
AK123846.1 | - | 4 | 486 | |||
AY026388.1 | - | 3 | 191 | |||
BC130304.1 | - | 3 | 191 | 40146335 | ||
BC144189.1 | - | 3 | 191 | 9052709 | ||
BX346769.2 | - | 3 | 714 |