Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297592.1 | NP_001284521.1 | 15 | 2329 | ||
NM_001297594.1 | NP_001284523.1 | 15 | 2351 | |||
NM_015187.4 | NP_056002.2 | 15 | 2546 | |||
XM_011513819.2 | XP_011512121.2 | 15 | 2540 | |||
XM_017007923.1 | XP_016863412.1 | 14 | 2184 | |||
GenBank mRNA | AB018289.1 | - | 14 | 2044 | ||
AK025282.1 | - | 3 | 212 | |||
AK289687.1 | - | 15 | 2294 | |||
AK300219.1 | - | 6 | 644 | |||
BC009945.2 | - | 8 | 1061 | |||
BC029422.1 | - | 5 | 471 | |||
BC041046.1 | - | 6 | 698 | |||
BC060867.1 | - | 15 | 2339 | 30344157 | ||
CR749817.1 | - | 15 | 2329 |