Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015202.3 | NP_056017.3 | 7 | 803 | ||
XM_005255201.3 | XP_005255258.1 | 8 | 951 | |||
XM_005255202.3 | XP_005255259.1 | 8 | 951 | |||
XM_005255203.3 | XP_005255260.1 | 7 | 822 | |||
XM_005255206.3 | XP_005255263.1 | 8 | 951 | |||
XM_006721025.3 | XP_006721088.1 | 6 | 690 | |||
XM_011545773.2 | XP_011544075.1 | 8 | 921 | |||
XM_011545774.1 | XP_011544076.1 | 7 | 867 | |||
XM_011545775.2 | XP_011544077.1 | 8 | 951 | |||
XM_011545776.2 | XP_011544078.1 | 7 | 819 | |||
XM_017023085.1 | XP_016878574.1 | 7 | 822 | |||
XM_017023086.1 | XP_016878575.1 | 6 | 756 | |||
XM_017023087.1 | XP_016878576.1 | 6 | 690 | |||
XM_017023088.1 | XP_016878577.1 | 4 | 584 | |||
GenBank mRNA | AB011128.2 | - | 7 | 801 | ||
AK097485.1 | - | 4 | 640 | |||
BC150276.1 | - | 7 | 801 | 9007250 |