Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286715.1 | NP_001273644.1 | 19 | 2910 | ||
NM_001286717.1 | NP_001273646.1 | 19 | 2790 | |||
NM_015239.2 | NP_056054.2 | 20 | 2855 | |||
XM_005251847.3 | XP_005251904.1 | 20 | 2921 | |||
XM_005251848.2 | XP_005251905.1 | 20 | 3099 | |||
XM_005251849.3 | XP_005251906.1 | 19 | 2845 | |||
XM_011518418.2 | XP_011516720.1 | 20 | 2921 | |||
XM_011518420.2 | XP_011516722.1 | 10 | 6174 | |||
XM_017014544.1 | XP_016870033.1 | 20 | 3466 | |||
XM_017014545.1 | XP_016870034.1 | 20 | 2801 | |||
XM_017014546.1 | XP_016870035.1 | 19 | 2688 | |||
XM_017014547.1 | XP_016870036.1 | 19 | 2845 | |||
XM_017014548.1 | XP_016870037.1 | 19 | 2788 | |||
XM_017014549.1 | XP_016870038.1 | 18 | 2807 | |||
GenBank mRNA | AB028958.2 | - | 20 | 2833 | ||
AK001544.1 | - | 21 | 2655 | |||
AK022562.1 | - | 8 | 1498 | |||
AK023280.1 | - | 8 | 1490 | |||
AK024698.1 | - | 4 | 440 | |||
AK295774.1 | - | 19 | 2790 | |||
AK299506.1 | - | 16 | 2350 | |||
AL833359.1 | - | 8 | 1447 | |||
BC060815.1 | - | 20 | 2820 | 30344122 | ||
BX648366.1 | - | 8 | 1902 |