Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
siRNA ID
HSS118624
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
133, 134
24527
NM_182961.3
NP_892006.3
134, 135
24905
XM_006715407.1
XP_006715470.1
134, 135
24818
XM_006715408.2
XP_006715471.1
134, 135
24810
XM_006715409.1
XP_006715472.1
133, 134
24797
XM_006715410.2
XP_006715473.1
134, 135
24822
XM_006715411.1
XP_006715474.1
133, 134
24767
XM_006715412.2
XP_006715475.1
134, 135
24807
XM_006715413.2
XP_006715476.1
134, 135
24822
XM_006715414.1
XP_006715477.1
132, 133
24746
XM_006715415.2
XP_006715478.1
134, 135
24822
XM_006715416.2
XP_006715479.1
134, 135
24807
XM_006715417.2
XP_006715480.1
133, 134
24681
XM_006715420.2
XP_006715483.1
133, 134
24669
XM_006715421.2
XP_006715484.1
133, 134
24666
XM_006715422.1
XP_006715485.1
133, 134
24659
XM_006715423.2
XP_006715486.1
134, 135
24822
XM_006715424.2
XP_006715487.1
134, 135
24822
XM_006715425.2
XP_006715488.1
134, 135
24822
XM_011535641.2
XP_011533943.1
134, 135
24819
XM_011535642.2
XP_011533944.1
134, 135
24807
XM_011535643.1
XP_011533945.1
133, 134
24653
XM_011535644.1
XP_011533946.1
120, 121
22869
XM_011535645.2
XP_011533947.1
116, 117
22262
XM_017010608.1
XP_016866097.1
135, 136
24937
XM_017010609.1
XP_016866098.1
134, 135
24744
XM_017010610.1
XP_016866099.1
133, 134
24723
XM_017010611.1
XP_016866100.1
133, 134
24526
XM_017010612.1
XP_016866101.1
132, 133
24450
XM_017010613.1
XP_016866102.1
134, 135
24819
XM_017010614.1
XP_016866103.1
133, 134
24666
XM_017010615.1
XP_016866104.1
133, 134
24666
XM_017010616.1
XP_016866105.1
134, 135
24822
XM_017010617.1
XP_016866106.1
134, 135
24819
XM_017010618.1
XP_016866107.1
134, 135
24807
XM_017010619.1
XP_016866108.1
120, 121
22901
GenBank mRNA
AB018339.2
-
22, 23
3542
AF444779.1
-
9, 10
1415
AF495910.1
-
134, 135
24905
AF535142.1
-
133, 134
24527
AK122691.1
-
14, 15
2185
AK127076.1
-
18, 19
2882
AK304825.1
-
8, 9
1107
AK316101.1
-
7, 8
1122
AY061755.1
-
49, 50
7995
AY061756.1
-
6, 7
985
AY184203.1
-
5, 6
850
AY184206.1
-
47, 48
7890
BC090927.1
-
7, 8
1167
BX537837.1
-
16, 17
2483
BX647837.1
-
19, 20
3077
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-xzndl:80/10.239.164.225:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0