Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBR4 SIRNA Assays ›
Gene Symbol:
UBR4
Gene Name:
ubiquitin protein ligase E3 component n-recognin 4
Gene Aliases:
RBAF600, ZUBR1, p600
Chromosome Location:
Chr.1: 19074506 - 19210334 on Build GRCh38
UniGene ID:
Hs.148078
Species:
Human
siRNA ID
HSS118633
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBR4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020765.2
NP_065816.2
16
2024
XM_011541108.2
XP_011539410.1
16
2106
XM_011541109.2
XP_011539411.1
16
2106
XM_011541110.2
XP_011539412.1
16
2106
XM_011541111.2
XP_011539413.1
16
2106
XM_011541112.2
XP_011539414.1
16
2106
XM_011541113.2
XP_011539415.1
16
2106
XM_011541114.2
XP_011539416.1
16
2106
XM_011541115.2
XP_011539417.1
16
2106
XM_011541116.2
XP_011539418.1
16
2106
XM_011541117.2
XP_011539419.1
16
2106
XM_011541118.2
XP_011539420.1
16
2106
XM_011541119.2
XP_011539421.1
16
2106
XM_011541120.2
XP_011539422.1
16
2106
XM_011541121.2
XP_011539423.1
16
2106
XM_017000822.1
XP_016856311.1
16
2106
XM_017000823.1
XP_016856312.1
16
2106
XM_017000824.1
XP_016856313.1
16
2106
XM_017000825.1
XP_016856314.1
16
2106
XM_017000826.1
XP_016856315.1
16
2106
XM_017000827.1
XP_016856316.1
16
2106
XM_017000828.1
XP_016856317.1
16
2106
XM_017000829.1
XP_016856318.1
16
2106
XM_017000830.1
XP_016856319.1
16
2106
GenBank mRNA
AF348492.1
-
16
2020
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
viral process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
calmodulin binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0