Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GPR161 SIRNA Assays ›
Gene Symbol:
GPR161
Gene Name:
G protein-coupled receptor 161
Gene Aliases:
RE2
Chromosome Location:
Chr.1: 168079542 - 168137667 on Build GRCh38
UniGene ID:
Hs.271809
Species:
Human
siRNA ID
HSS118759
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GPR161
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267609.1
NP_001254538.1
4
1710
NM_001267610.1
NP_001254539.1
4
1673
NM_001267611.1
NP_001254540.1
3
1260
NM_001267612.1
NP_001254541.1
3
1216
NM_001267613.1
NP_001254542.1
2
1109
NM_001267614.1
NP_001254543.1
2
842
NM_153832.2
NP_722561.1
5
1667
XM_005245055.2
XP_005245112.1
4
1634
XM_005245056.2
XP_005245113.1
3
1241
XM_005245057.4
XP_005245114.1
3
1258
XM_006711251.1
XP_006711314.1
5
1680
XM_006711253.2
XP_006711316.2
3
1473
XM_011509370.1
XP_011507672.1
5
1978
XM_011509371.2
XP_011507673.1
5
1424
XM_011509372.1
XP_011507674.1
5
1756
XM_011509373.2
XP_011507675.1
5
1682
XM_011509375.2
XP_011507677.1
5
1468
XM_011509376.2
XP_011507678.1
4
1599
XM_011509377.1
XP_011507679.1
3
1555
XM_011509378.1
XP_011507680.1
3
1527
XM_017000858.1
XP_016856347.1
4
2127
XM_017000859.1
XP_016856348.1
3
2081
XM_017000860.1
XP_016856349.1
4
2175
XM_017000861.1
XP_016856350.1
5
2221
XM_017000862.1
XP_016856351.1
5
1804
GenBank mRNA
AF091890.1
-
2
1092
AK056040.1
-
3
1163
AK122656.1
-
4
1589
AK298797.1
-
2
1067
AK298850.1
-
2
842
AK299634.1
-
4
1341
AK310019.1
-
3
1177
AY275468.1
-
2
1032
BC028163.1
-
5
1346
5246359
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
multicellular organism development
positive regulation of cAMP biosynthetic process
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
G-protein coupled receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0