Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014296.2 | NP_055111.1 | 7 | 1015 | ||
XM_011533555.2 | XP_011531857.1 | 7 | 866 | |||
XM_011533556.2 | XP_011531858.1 | 8 | 1098 | |||
XM_011533557.2 | XP_011531859.1 | 7 | 988 | |||
XM_017006043.1 | XP_016861532.1 | 8 | 1098 | |||
XM_017006044.1 | XP_016861533.1 | 7 | 988 | |||
XM_017006045.1 | XP_016861534.1 | 8 | 1054 | |||
GenBank mRNA | AB028639.1 | - | 7 | 996 | ||
AK314253.1 | - | 7 | 977 | |||
BC056202.1 | - | 7 | 993 | 5274889 |