Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201427.1 | NP_001188356.1 | 10 | 1219 | ||
NM_015345.3 | NP_056160.2 | 10 | 1283 | |||
XM_006715039.3 | XP_006715102.3 | 10 | 1864 | |||
XM_006715040.3 | XP_006715103.3 | 10 | 1863 | |||
XM_006715042.2 | XP_006715105.1 | 10 | 3859 | |||
XM_006715043.2 | XP_006715106.1 | 10 | 1236 | |||
XM_006715045.3 | XP_006715108.1 | 10 | 2451 | |||
XM_006715046.3 | XP_006715109.1 | 10 | 1629 | |||
XM_017010630.1 | XP_016866119.1 | 10 | 2475 | |||
GenBank mRNA | AB002379.2 | - | 10 | 1219 | ||
AK310941.1 | - | 10 | 1226 | |||
AL833083.2 | - | 10 | 1263 |