Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012343.3 | NP_036475.3 | 16, 17 | 2719 | ||
NM_182977.2 | NP_892022.2 | 16, 17 | 2685 | |||
XM_005248274.4 | XP_005248331.1 | 16, 17 | 2697 | |||
XM_005248275.4 | XP_005248332.1 | 11, 12 | 1947 | |||
XM_006714461.3 | XP_006714524.1 | 15, 16 | 2311 | |||
XM_011514001.2 | XP_011512303.1 | 16, 17 | 2716 | |||
XM_011514002.2 | XP_011512304.1 | 15, 16 | 2406 | |||
XM_017009293.1 | XP_016864782.1 | 16, 17 | 2587 | |||
GenBank mRNA | AK297558.1 | - | 9, 10 | 1332 | ||
AK299622.1 | - | 8, 9 | 1149 | |||
AL831822.2 | - | 16, 17 | 2686 | |||
BC110543.1 | - | 16, 17 | 2535 | 40036686 | ||
BC110544.1 | - | 15, 16 | 2532 | 40036689 | ||
U40490.1 | - | 16, 17 | 2607 |