Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX2 SIRNA Assays ›
Gene Symbol:
RBFOX2
Gene Name:
RNA binding protein, fox-1 homolog 2
Gene Aliases:
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
Chromosome Location:
Chr.22: 35738736 - 36028892 on Build GRCh38
UniGene ID:
Hs.282998
Species:
Human
siRNA ID
HSS118926
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBFOX2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031695.2
NP_001026865.1
7
1003
NM_001082576.1
NP_001076045.1
7
1003
NM_001082577.1
NP_001076046.1
7
1003
NM_001082578.1
NP_001076047.1
8
963
NM_001082579.1
NP_001076048.1
8
960
NM_014309.2
NP_055124.1
7
1006
XM_005261428.2
XP_005261485.1
8
1318
XM_005261429.2
XP_005261486.1
8
1315
XM_005261430.2
XP_005261487.1
8
1318
XM_005261431.2
XP_005261488.1
8
1318
XM_005261432.2
XP_005261489.1
8
1315
XM_005261433.2
XP_005261490.1
8
1318
XM_005261435.1
XP_005261492.1
7
1007
XM_005261437.1
XP_005261494.1
7
1010
XM_006724185.2
XP_006724248.1
8
1318
XM_006724186.2
XP_006724249.1
8
1315
XM_006724187.2
XP_006724250.1
8
1318
XM_006724188.2
XP_006724251.1
8
1318
XM_006724189.3
XP_006724252.1
8
1011
XM_006724190.1
XP_006724253.1
8
705
XM_006724191.1
XP_006724254.1
7
1010
XM_006724192.2
XP_006724255.1
7
840
XM_006724193.3
XP_006724256.1
8
1318
XM_006724194.2
XP_006724257.1
7
782
XM_017028686.1
XP_016884175.1
8
697
XM_017028687.1
XP_016884176.1
8
878
XM_017028688.1
XP_016884177.1
8
1315
XM_017028689.1
XP_016884178.1
8
783
XM_017028690.1
XP_016884179.1
7
760
XM_017028691.1
XP_016884180.1
7
757
XM_017028692.1
XP_016884181.1
8
1052
XM_017028693.1
XP_016884182.1
7
1010
XM_017028694.1
XP_016884183.1
7
1007
XM_017028695.1
XP_016884184.1
8
767
XM_017028696.1
XP_016884185.1
7
1010
XM_017028697.1
XP_016884186.1
8
1787
XM_017028698.1
XP_016884187.1
7
1007
GenBank mRNA
AB649123.1
-
7
638
AF229058.1
-
7
808
AK055213.1
-
8
924
AK291460.1
-
7
887
AK314632.1
-
7
822
AL009266.1
-
7
811
AM419009.1
-
7
662
AY072786.1
-
8
773
AY960684.1
-
7
638
BC013115.1
-
6
773
4346741
BC025281.1
-
7
826
4555865
CR456559.1
-
7
704
DQ778625.1
-
8
851
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
fibroblast growth factor receptor signaling pathway
regulation of definitive erythrocyte differentiation
RNA metabolic process
radial glia guided migration of Purkinje cell
intracellular estrogen receptor signaling pathway
regulation of cell proliferation
negative regulation of transcription, DNA-templated
dendrite morphogenesis
neuromuscular process controlling balance
nucleotide binding
transcription corepressor activity
RNA binding
mRNA binding
protein binding
transcription factor binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline