Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319116.1 | NP_001306045.1 | 14, 15 | 1523 | ||
NM_001319117.1 | NP_001306046.1 | 13, 14 | 1415 | |||
NM_001319118.1 | NP_001306047.1 | 14, 15 | 1451 | |||
NM_001319119.1 | NP_001306048.1 | 14, 15 | 1731 | |||
NM_001319120.1 | NP_001306049.1 | 13, 14 | 1440 | |||
NM_001319121.1 | NP_001306050.1 | 15, 16 | 1424 | |||
NM_001319122.1 | NP_001306051.1 | 14, 15 | 1516 | |||
NM_012100.3 | NP_036232.2 | 14, 15 | 1548 | |||
NR_134970.1 | 12, 13 | 1364 | ||||
GenBank mRNA | AF005050.2 | - | 14, 15 | 1523 | ||
AK001777.1 | - | 14, 15 | 1547 | |||
AK001977.1 | - | 14, 15 | 1520 | |||
AK023005.1 | - | 14, 15 | 1462 | |||
AK301919.1 | - | 14, 15 | 1508 | |||
BC000653.2 | - | 14, 15 | 1430 | 3343505 | ||
BC004854.2 | - | 10, 11 | 1081 |