Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF346 SIRNA Assays ›
Gene Symbol:
ZNF346
Gene Name:
zinc finger protein 346
Gene Aliases:
JAZ, Zfp346
Chromosome Location:
Chr.5: 177022680 - 177081189 on Build GRCh38
UniGene ID:
Hs.484259
Species:
Human
siRNA ID
HSS118977
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF346
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308213.1
NP_001295142.1
8
969
NM_001308214.1
NP_001295143.1
6
798
NM_001308215.1
NP_001295144.1
6
708
NM_001308216.1
NP_001295145.1
4
469
NM_001308218.1
NP_001295147.1
4
600
NM_001308219.1
NP_001295148.1
4
469
NM_001308221.1
NP_001295150.1
3
366
NM_001308223.1
NP_001295152.1
3
366
NM_012279.3
NP_036411.1
7
894
NR_131773.1
6
749
NR_131774.1
5
563
XM_005265860.2
XP_005265917.1
8
962
XM_005265864.3
XP_005265921.1
7
728
XM_011534493.1
XP_011532795.1
8
962
XM_011534494.1
XP_011532796.1
7
868
XM_011534495.2
XP_011532797.1
7
868
XM_011534496.2
XP_011532798.1
7
820
XM_017009294.1
XP_016864783.1
7
942
XM_017009295.1
XP_016864784.1
7
894
XM_017009296.1
XP_016864785.1
7
894
XM_017009297.1
XP_016864786.1
6
848
XM_017009299.1
XP_016864788.1
6
800
XM_017009300.1
XP_016864789.1
7
1232
XM_017009301.1
XP_016864790.1
6
708
XM_017009302.1
XP_016864791.1
8
907
XM_017009303.1
XP_016864792.1
5
1005
GenBank mRNA
AF083340.1
-
7
867
AK026627.1
-
7
862
AK131469.1
-
8
914
AK295549.1
-
6
774
AK297448.1
-
4
584
AK297584.1
-
3
350
AK300065.1
-
6
713
AK300738.1
-
5
508
AL359576.1
-
7
842
BC007775.2
-
7
849
4301820
BX331331.2
-
4
424
CR749692.1
-
7
842
DA169130.1
-
4
442
DR003225.1
-
3
369
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of apoptotic process
intrinsic apoptotic signaling pathway by p53 class mediator
p53 binding
double-stranded RNA binding
protein binding
zinc ion binding
enzyme binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0