Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145125.1 | NP_001138597.1 | 5 | 1032 | ||
NM_001291663.1 | NP_001278592.1 | 7 | 1194 | |||
NM_013446.3 | NP_038474.2 | 5 | 1032 | |||
NR_117084.1 | 3 | 575 | ||||
XM_011515996.1 | XP_011514298.1 | 4 | 821 | |||
XM_011515997.2 | XP_011514299.1 | 5 | 1089 | |||
XM_011515998.1 | XP_011514300.1 | 5 | 774 | |||
GenBank mRNA | AB209298.1 | - | 5 | 799 | ||
AF192784.1 | - | 5 | 927 | |||
AK027137.1 | - | 4 | 544 | |||
AK127030.1 | - | 6 | 1649 | |||
AK297361.1 | - | 3 | 374 | |||
AK315552.1 | - | 5 | 831 | |||
AL136812.1 | - | 5 | 930 | |||
AM236048.1 | - | 5 | 813 | |||
BC025955.1 | - | 5 | 965 | 4869484 | ||
BC037400.1 | - | 5 | 817 | 5176176 | ||
BC064838.1 | - | 5 | 810 | 6173715 | ||
CA438497.1 | - | 1 | 651 |