Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271707.1 | NP_001258636.1 | 6, 7 | 1121 | ||
NM_014160.4 | NP_054879.3 | 7, 8 | 1250 | |||
GenBank mRNA | AF302084.1 | - | 7, 8 | 1164 | ||
AK091318.1 | - | 7, 8 | 1195 | |||
AK298463.1 | - | 6, 7 | 1048 | |||
AL834512.1 | - | 4, 5 | 772 | |||
BC001799.2 | - | 5, 6 | 959 | |||
BC015715.1 | - | 7, 8 | 1192 | 3916199 |