Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130852.1 | NP_001124324.1 | 6 | 962 | ||
NM_012116.3 | NP_036248.3 | 7 | 1100 | |||
XM_005258696.3 | XP_005258753.1 | 7 | 1107 | |||
XM_011526688.2 | XP_011524990.1 | 7 | 1108 | |||
XM_011526689.2 | XP_011524991.1 | 6 | 972 | |||
XM_011526690.2 | XP_011524992.1 | 6 | 1008 | |||
GenBank mRNA | AB028645.1 | - | 7 | 1078 | ||
AF117646.1 | - | 7 | 1100 | |||
AF117647.1 | - | 6 | 962 | |||
BC028915.1 | - | 7 | 1058 | 4777669 |