Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195797.1 | NP_001182726.1 | 4 | 490 | ||
NM_015364.4 | NP_056179.3 | 5 | 580 | |||
GenBank mRNA | AB018549.1 | - | 5 | 591 | ||
AB446498.1 | - | 5 | 466 | |||
AF168121.1 | - | 5 | 487 | |||
BC020690.1 | - | 5 | 490 | 4767246 |