Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012261.3 | NP_036393.1 | 2, 3 | 723 | ||
GenBank mRNA | AL121740.1 | - | 2, 3 | 430 | ||
BC050727.1 | - | 2, 3 | 426 | 6044324 |