Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015380.4 | NP_056195.3 | 5 | 538 | ||
GenBank mRNA | AF151809.1 | - | 5 | 450 | ||
AK001087.1 | - | 5 | 477 | |||
AK222657.1 | - | 5 | 412 | |||
AK312773.1 | - | 5 | 373 | |||
AY189688.1 | - | 5 | 453 | |||
BC007830.1 | - | 5 | 445 | 3635175 | ||
BC011681.2 | - | 5 | 498 | 3010765 | ||
BC015200.1 | - | 5 | 388 | 4333051 | ||
CR456483.1 | - | 5 | 350 | |||
DB030637.1 | - | 5 | 538 |