Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267041.2 | NP_001253970.1 | 10 | 1161 | ||
NM_001282342.1 | NP_001269271.1 | 9 | 1084 | |||
NM_014154.4 | NP_054873.2 | 12 | 1330 | |||
NM_015396.5 | NP_056211.2 | 12 | 1330 | |||
NM_213654.3 | NP_998819.1 | 12 | 1384 | |||
XM_006713564.3 | XP_006713627.1 | 11 | 1249 | |||
XM_006713565.2 | XP_006713628.1 | 12 | 1290 | |||
XM_006713566.3 | XP_006713629.1 | 10 | 1144 | |||
XM_006713567.3 | XP_006713630.1 | 11 | 1249 | |||
XM_011512632.1 | XP_011510934.1 | 11 | 1197 | |||
XM_011512633.1 | XP_011510935.1 | 11 | 1251 | |||
XM_017006096.1 | XP_016861585.1 | 10 | 1144 | |||
XM_017006100.1 | XP_016861589.1 | 11 | 1249 | |||
XM_017006101.1 | XP_016861590.1 | 11 | 1159 | |||
GenBank mRNA | AF109184.1 | - | 12 | 1083 | ||
AF161541.1 | - | 12 | 1048 | |||
AK001721.1 | - | 10 | 892 | |||
AK001882.1 | - | 3 | 465 | |||
AK098500.1 | - | 2 | 1619 | |||
AK289577.1 | - | 12 | 1026 | |||
AK296837.1 | - | 10 | 861 | |||
AK299229.1 | - | 6 | 522 | |||
AK299769.1 | - | 9 | 804 | |||
AK304558.1 | - | 10 | 837 | |||
AK315932.1 | - | 6 | 527 | |||
BC013424.1 | - | 12 | 1084 | 3892143 | ||
BC032661.1 | - | 12 | 1194 | 5530425 | ||
BC041699.1 | - | 12 | 1009 | 5240423 |