Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCHY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009922.2 | NP_001009922.1 | 4 | 528 | ||
NM_001278536.1 | NP_001265465.1 | 3 | 408 | |||
NM_001278537.1 | NP_001265466.1 | 3 | 408 | |||
NM_001278538.1 | NP_001265467.1 | 4 | 484 | |||
NM_001278539.1 | NP_001265468.1 | 4 | 566 | |||
NM_015436.3 | NP_056251.2 | 4 | 528 | |||
NR_037913.1 | 4 | 528 | ||||
NR_037914.1 | 4 | 524 | ||||
NR_103723.1 | 4 | 528 | ||||
NR_103724.1 | 4 | 528 | ||||
NR_103725.1 | 3 | 302 | ||||
XM_011531838.2 | XP_011530140.1 | 4 | 497 | |||
XM_011531839.2 | XP_011530141.1 | 4 | 685 | |||
GenBank mRNA | AB209072.1 | - | 4 | 391 | ||
AF247041.1 | - | 4 | 384 | |||
AF255666.1 | - | 4 | 410 | |||
AF305424.1 | - | 4 | 418 | |||
AK091501.1 | - | 4 | 499 | |||
AK308754.1 | - | 3 | 261 | |||
AK308848.1 | - | 1 | 310 | |||
AK309555.1 | - | 4 | 371 | |||
AL050144.1 | - | 3 | 287 | |||
AY888047.1 | - | 4 | 410 | |||
BC047393.1 | - | 4 | 528 | 5301408 | ||
BF102905.1 | - | 3 | 302 | |||
BG703058.1 | - | 4 | 397 | |||
BI913695.1 | - | 3 | 221 | |||
BP314571.1 | - | 3 | 275 | |||
BQ959587.1 | - | 4 | 357 | |||
CF597260.1 | - | 2 | 99 | |||
DA197338.1 | - | 4 | 484 | |||
GQ250944.1 | - | 4 | 384 | |||
GQ250945.1 | - | 4 | 384 | |||
GQ250946.1 | - | 4 | 384 | |||
GU937000.1 | - | 4 | 412 | |||
JF691538.1 | - | 4 | 384 | |||
JF691539.1 | - | 4 | 384 |