Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042549.1 | NP_001036014.1 | 1 | 175 | ||
NM_001297736.1 | NP_001284665.1 | 1 | 175 | |||
NM_001297737.1 | NP_001284666.1 | 1 | 175 | |||
NM_001297739.1 | NP_001284668.1 | 1 | 175 | |||
NM_015471.3 | NP_056286.3 | 1 | 175 | |||
GenBank mRNA | AF201941.1 | - | 1 | 147 | ||
AF255793.1 | - | 1 | 161 | |||
AK222996.1 | - | 1 | 145 | |||
AK223260.1 | - | 1 | 155 | |||
AK303250.1 | - | 1 | 155 | |||
AK313730.1 | - | 1 | 171 | |||
BC007067.1 | - | 1 | 149 | 3997280 | ||
BC036048.1 | - | 1 | 153 | |||
BU570983.1 | - | 1 | 155 | |||
CK003923.1 | - | 1 | 158 | |||
DB045932.1 | - | 1 | 175 |