Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015474.3 | NP_056289.2 | 4 | 609 | ||
XM_005260384.3 | XP_005260441.1 | 4 | 582 | |||
XM_011528761.2 | XP_011527063.1 | 4 | 582 | |||
GenBank mRNA | AB013847.1 | - | 4 | 409 | ||
AB208944.1 | - | 4 | 439 | |||
AF228421.1 | - | 4 | 562 | |||
AK027811.1 | - | 4 | 582 | |||
AK304795.1 | - | 5 | 537 | |||
AK315169.1 | - | 4 | 542 | |||
AL050267.1 | - | 4 | 484 | |||
BC036450.2 | - | 4 | 437 | 4816438 |