Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015483.2 | NP_056298.2 | 4 | 1828 | ||
XM_005249696.2 | XP_005249753.1 | 4 | 1684 | |||
XM_005249697.2 | XP_005249754.1 | 4 | 1417 | |||
XM_011515256.2 | XP_011513558.1 | 4 | 1563 | |||
XM_011515257.2 | XP_011513559.1 | 4 | 1297 | |||
GenBank mRNA | AB040922.1 | - | 1 | 2452 | ||
AF161402.1 | - | 1 | 582 | |||
AK024557.1 | - | 1 | 793 | |||
AK292802.1 | - | 4 | 1550 | |||
AL117562.1 | - | 1 | 346 | |||
BC037887.2 | - | 4 | 1732 | 5274706 | ||
BC047107.1 | - | 4 | 1565 | 5275217 |