Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283014.1 | NP_001269943.1 | 7 | 979 | ||
NM_001283015.1 | NP_001269944.1 | 8 | 1134 | |||
NM_001283016.1 | NP_001269945.1 | 10 | 1318 | |||
NM_001283017.1 | NP_001269946.1 | 9 | 1211 | |||
NM_001283019.1 | NP_001269948.1 | 8 | 1139 | |||
NM_015901.5 | NP_056985.3 | 9 | 1246 | |||
NR_104264.1 | 9 | 1284 | ||||
GenBank mRNA | AK300980.1 | - | 10 | 1171 | ||
AK303235.1 | - | 8 | 1015 | |||
AK316538.1 | - | 9 | 1064 | |||
AL050114.1 | - | 6 | 806 | |||
BC031224.1 | - | 9 | 1204 | 4824935 | ||
BC046173.2 | - | 7 | 832 | 3873776 |