Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167890.1 | NP_001161362.1 | 8 | 1130 | ||
NM_015507.3 | NP_056322.2 | 8 | 1130 | |||
GenBank mRNA | AF186084.1 | - | 8 | 988 | ||
AF193055.1 | - | 8 | 1055 | |||
AJ245671.1 | - | 8 | 1083 | |||
AK075214.1 | - | 8 | 1100 | |||
AK300137.1 | - | 8 | 866 | |||
AK315018.1 | - | 8 | 968 | |||
AY358333.1 | - | 8 | 977 | |||
BC038587.1 | - | 8 | 939 | 5767603 |