Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167890.1 | NP_001161362.1 | 2, 3 | 471 | ||
NM_015507.3 | NP_056322.2 | 2, 3 | 471 | |||
GenBank mRNA | AF186084.1 | - | 2, 3 | 329 | ||
AF193055.1 | - | 2, 3 | 397 | |||
AJ245671.1 | - | 2, 3 | 424 | |||
AK075214.1 | - | 2, 3 | 441 | |||
AK300137.1 | - | 2, 3 | 207 | |||
AK300236.1 | - | 2, 3 | 441 | |||
AK315018.1 | - | 2, 3 | 309 | |||
AL117610.1 | - | 2, 3 | 201 | |||
AY358333.1 | - | 2, 3 | 318 | |||
BC038587.1 | - | 2, 3 | 280 | 5767603 | ||
DA534733.1 | - | 2, 3 | 442 | |||
DC380958.1 | - | 2, 3 | 471 |