Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPCEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130699.1 | NP_001124171.1 | 10 | 719 | ||
NM_001130700.1 | NP_001124172.1 | 9 | 679 | |||
NM_015553.2 | NP_056368.1 | 9 | 676 | |||
XM_005266919.4 | XP_005266976.1 | 10 | 836 | |||
XM_005266921.4 | XP_005266978.1 | 9 | 762 | |||
XM_005266923.3 | XP_005266980.1 | 10 | 770 | |||
XM_011535742.2 | XP_011534044.1 | 10 | 834 | |||
XM_011535744.2 | XP_011534046.1 | 6 | 1189 | |||
XM_017010716.1 | XP_016866205.1 | 10 | 806 | |||
XM_017010717.1 | XP_016866206.1 | 9 | 920 | |||
XM_017010718.1 | XP_016866207.1 | 9 | 917 | |||
GenBank mRNA | AB007863.1 | - | 5 | 457 | ||
AJ310566.1 | - | 9 | 677 | |||
AK093997.1 | - | 1 | 547 | |||
AK289917.1 | - | 9 | 648 | |||
BC105015.1 | - | 8 | 585 | 8144018 | ||
BC105043.1 | - | 8 | 585 | 8144046 | ||
BC143623.1 | - | 8 | 588 | 9052136 | ||
BC143629.1 | - | 8 | 585 | 9052142 | ||
BX647254.1 | - | 10 | 719 |