Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015557.2 | NP_056372.1 | 22 | 3395 | ||
GenBank mRNA | AB007913.1 | - | 4 | 356 | ||
AF425231.1 | - | 22 | 3384 | |||
AK091386.1 | - | 13 | 1871 | |||
AK126012.1 | - | 2 | 131 | |||
AK127046.1 | - | 22 | 3284 | |||
AL110281.1 | - | 11 | 1432 | |||
AL117491.1 | - | 12 | 1588 | |||
AL137693.1 | - | 11 | 1411 |