Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAI14 SIRNA Assays ›
Gene Symbol:
RAI14
Gene Name:
retinoic acid induced 14
Gene Aliases:
NORPEG, RAI13
Chromosome Location:
Chr.5: 34656328 - 34832612 on Build GRCh38
UniGene ID:
Hs.431400
Species:
Human
siRNA ID
HSS119767
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RAI14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145520.1
NP_001138992.1
15
1405
NM_001145521.1
NP_001138993.1
16
1518
NM_001145522.1
NP_001138994.1
14
1260
NM_001145523.1
NP_001138995.1
14
1279
NM_001145525.1
NP_001138997.1
17
1699
NM_015577.2
NP_056392.2
15
1347
XM_006714469.2
XP_006714532.1
15
1314
XM_011514016.2
XP_011512318.2
14
1261
XM_011514017.2
XP_011512319.1
15
1438
XM_011514018.1
XP_011512320.1
15
1439
XM_011514019.1
XP_011512321.1
16
1413
XM_011514020.1
XP_011512322.1
15
1372
XM_011514021.1
XP_011512323.1
15
1276
XM_011514022.1
XP_011512324.1
15
1255
XM_011514023.1
XP_011512325.1
15
1297
XM_011514024.1
XP_011512326.1
15
1290
XM_011514025.2
XP_011512327.2
13
1171
XM_017009334.1
XP_016864823.1
14
1456
XM_017009335.1
XP_016864824.1
14
1339
XM_017009336.1
XP_016864825.1
14
1203
GenBank mRNA
AB037755.1
-
15
1405
AF155135.1
-
15
1262
AK297964.1
-
8
697
AK308455.1
-
6
374
AK314379.1
-
14
1149
AY317139.1
-
17
1652
AY354204.2
-
14
1279
BC052988.1
-
15
1263
6164087
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0