Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IFT172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015662.2 | NP_056477.1 | 25 | 2847 | ||
XM_005264254.1 | XP_005264311.1 | 25 | 2678 | |||
XM_006711986.3 | XP_006712049.1 | 26 | 2939 | |||
XM_006711987.1 | XP_006712050.1 | 25 | 2744 | |||
XM_011532757.2 | XP_011531059.1 | 25 | 3053 | |||
XM_011532758.1 | XP_011531060.1 | 25 | 2744 | |||
XM_011532759.2 | XP_011531061.1 | 11 | 1329 | |||
XM_011532760.2 | XP_011531062.1 | 8 | 1009 | |||
XM_017003790.1 | XP_016859279.1 | 26 | 2873 | |||
XM_017003791.1 | XP_016859280.1 | 24 | 2814 | |||
XM_017003792.1 | XP_016859281.1 | 25 | 2744 | |||
XM_017003793.1 | XP_016859282.1 | 21 | 2254 | |||
XM_017003794.1 | XP_016859283.1 | 19 | 1957 | |||
XM_017003795.1 | XP_016859284.1 | 23 | 2806 | |||
GenBank mRNA | AB033005.1 | - | 7 | 712 | ||
AL110218.1 | - | 22 | 2403 | |||
BC137126.1 | - | 25 | 2737 | 9021123 |