Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039592.1 | NP_001034681.1 | 2 | 325 | ||
NM_172312.1 | NP_758516.1 | 2 | 325 | |||
GenBank mRNA | AK302313.1 | - | 2 | 233 | ||
BC019247.1 | - | 2 | 230 | 4825175 | ||
BI757195.1 | - | 2 | 312 | |||
S83157.1 | - | 2 | 326 | |||
U12978.2 | - | 2 | 326 |