Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012369.2 | NP_036501.2 | 1 | 950 | ||
XM_011516019.2 | XP_011514321.1 | 4 | 1496 | |||
XM_011516020.2 | XP_011514322.1 | 4 | 1271 | |||
GenBank mRNA | BC104975.1 | - | 1 | 950 | 8143978 |