Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099274.1 | NP_001092744.1 | 6 | 1262 | ||
NM_012461.2 | NP_036593.2 | 6 | 1262 | |||
XM_005267529.3 | XP_005267586.1 | 5 | 1088 | |||
XM_011536642.2 | XP_011534944.1 | 5 | 1157 | |||
XM_017021216.1 | XP_016876705.1 | 2 | 794 | |||
XM_017021217.1 | XP_016876706.1 | 2 | 386 | |||
GenBank mRNA | AF195512.1 | - | 6 | 1182 | ||
AK023166.1 | - | 4 | 1653 | |||
AK225379.1 | - | 6 | 1206 | |||
AK294119.1 | - | 5 | 1088 | |||
AK315084.1 | - | 6 | 1150 | |||
BC005030.1 | - | 6 | 1043 | 3690254 | ||
BC019343.1 | - | 6 | 1220 | 3505009 | ||
BX161478.1 | - | 4 | 990 | |||
EU851975.1 | - | 3 | 436 |