Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012110.3 | NP_036242.1 | 4, 5 | 732 | ||
XM_006714037.3 | XP_006714100.1 | 4, 5 | 648 | |||
GenBank mRNA | AF159423.1 | - | 4, 5 | 633 | ||
AK312427.1 | - | 4, 5 | 695 | |||
BC034691.1 | - | 4, 5 | 644 | 4747392 |