Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012430.4 | NP_036562.2 | 7 | 835 | ||
XM_011512673.2 | XP_011510975.1 | 7 | 2351 | |||
XM_011512675.2 | XP_011510977.1 | 8 | 1002 | |||
XM_011512676.2 | XP_011510978.1 | 7 | 891 | |||
GenBank mRNA | AF100749.1 | - | 7 | 817 | ||
AK057587.1 | - | 6 | 1651 | |||
AK315771.1 | - | 7 | 804 | |||
BC007122.1 | - | 6 | 783 | 3459649 |