Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPK1 SIRNA Assays ›
Gene Symbol:
TPK1
Gene Name:
thiamin pyrophosphokinase 1
Gene Aliases:
HTPK1, PP20, THMD5
Chromosome Location:
Chr.7: 144451941 - 144836424 on Build GRCh38
UniGene ID:
Hs.660232
Species:
Human
siRNA ID
HSS120278
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042482.1
NP_001035947.1
6
404
NM_022445.3
NP_071890.2
6
404
XM_005249970.1
XP_005250027.1
6
382
XM_011516031.1
XP_011514333.1
7
483
XM_011516032.2
XP_011514334.1
7
619
XM_011516033.2
XP_011514335.1
7
489
XM_011516034.2
XP_011514336.1
7
546
XM_011516035.2
XP_011514337.1
7
483
XM_011516037.2
XP_011514339.1
6
730
XM_011516039.2
XP_011514341.1
6
665
XM_011516040.2
XP_011514342.1
7
483
XM_011516041.2
XP_011514343.1
5
633
XM_011516043.1
XP_011514345.1
7
483
XM_011516044.1
XP_011514346.1
4
307
XM_011516046.1
XP_011514348.1
7
483
XM_011516047.2
XP_011514349.1
3
1154
XM_011516048.1
XP_011514350.1
2
120
XM_011516050.1
XP_011514352.1
4
213
XM_017011969.1
XP_016867458.1
7
483
XM_017011970.1
XP_016867459.1
6
457
XM_017011971.1
XP_016867460.1
6
407
XM_017011972.1
XP_016867461.1
7
483
XM_017011973.1
XP_016867462.1
6
549
XM_017011974.1
XP_016867463.1
6
394
XM_017011975.1
XP_016867464.1
6
345
XM_017011976.1
XP_016867465.1
6
404
XM_017011977.1
XP_016867466.1
7
497
XM_017011978.1
XP_016867467.1
6
456
XM_017011979.1
XP_016867468.1
6
536
XM_017011980.1
XP_016867469.1
3
242
XM_017011981.1
XP_016867470.1
2
336
XM_017011982.1
XP_016867471.1
4
303
XM_017011983.1
XP_016867472.1
4
247
XM_017011984.1
XP_016867473.1
4
306
XM_017011985.1
XP_016867474.1
4
262
XM_017011986.1
XP_016867475.1
4
268
GenBank mRNA
AB028138.1
-
5
319
AF297710.1
-
6
391
AK026374.1
-
4
211
AK091736.1
-
6
343
AK128661.1
-
5
357
AK289652.1
-
6
379
AK308693.1
-
6
404
AY206415.1
-
6
386
BC040555.1
-
7
552
5269277
BC068460.1
-
6
383
5288626
BI770816.1
-
5
318
BX446289.2
-
6
385
CR749248.1
-
4
391
DA135998.1
-
6
404
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
thiamine metabolic process
thiamine diphosphate biosynthetic process
phosphorylation
thiamine-containing compound metabolic process
thiamine diphosphokinase activity
ATP binding
kinase activity
thiamine binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0